Assay Details
Gene Symbol: Il22ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178258.5 | NP_839989.2 | 2 - 3 | 207 | 114 | |
GenBank mRNA | AF493604.1 | - | 2 - 3 | 195 | 114 | |
AJ555484.1 | - | 2 - 3 | 201 | 114 | ||
AK138913.1 | - | 2 - 3 | 208 | 114 | ||
AK139024.1 | - | 2 - 3 | 238 | 114 | ||
BC124998.1 | - | 2 - 3 | 197 | 40044741 | 114 | |
BC126872.1 | - | 2 - 3 | 207 | 40044756 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
