Assay Details
Gene Symbol: Il22ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178258.5 | NP_839989.2 | 3 - 4 | 384 | 60 | |
GenBank mRNA | AF493604.1 | - | 3 - 4 | 372 | 60 | |
AJ555484.1 | - | 3 - 4 | 378 | 60 | ||
AK040641.1 | - | 3 - 4 | 393 | 60 | ||
AK138913.1 | - | 3 - 4 | 385 | 60 | ||
AK139024.1 | - | 3 - 4 | 415 | 60 | ||
BC124998.1 | - | 3 - 4 | 374 | 40044741 | 60 | |
BC126872.1 | - | 3 - 4 | 384 | 40044756 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046064 | AK040641.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046064 | AK138913.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046064 | NM_178258.5 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 944668 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 110906 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389046 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 896793 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 205221 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819613 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 4539 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 149834 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118315 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144966 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046064 | AK040641.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046064 | AK138913.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046064 | NM_178258.5 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P659290 | AK139024.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P659290 | BC124998.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P659290 | BC126872.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P659290 | AK040641.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P659290 | AK138913.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982156 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982172 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982197 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982151 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982194 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982195 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982155 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982168 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982169 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982180 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982185 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982196 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982167 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982176 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982164 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982170 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982177 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982186 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982162 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982160 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982182 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982191 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982152 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982198 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982174 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982159 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26982165 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144966 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118315 | - | - |
Whole Mouse Genome G4122A | A_52_P1004997 | AK040641.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P659290 | AK139024.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P659290 | BC124998.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P659290 | BC126872.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P659290 | AK040641.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P659290 | AK138913.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131490 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118315 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144966 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08422274 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08777195 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08339594 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08777198 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08777197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982209 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26982270 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982161 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982155 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982234 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982216 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982202 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08448062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982157 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982199 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982177 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982195 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982176 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982185 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982164 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982218 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982149 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982174 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982217 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982213 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982236 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982209 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08422958 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08422945 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08267725 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982140 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982159 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982167 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982178 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982205 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08357168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982191 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982151 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982221 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982227 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982203 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982143 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982207 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982156 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982182 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982170 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982160 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982186 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982189 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982183 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982139 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982215 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982237 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982196 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08357172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982210 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08339594 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08267726 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982152 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26982201 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08721925 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10361912 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10361908 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10361909 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10361907 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10361911 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10361913 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10361910 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1437665_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10361906 | - | - |
Mouse Genome 430 2.0 Array | 1437665_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1437665_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237310_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237310_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1000001001.mm.1 | TC1000000143.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000001001.mm.1 | NM_178258.5.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000001001.mm.1 | NM_178258.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000000999.mm.1 | TC1000000143.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000000999.mm.1 | NM_178258.5.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000000999.mm.1 | NM_178258.5 | Refseq |
Affymetrix HT Mouse Genome 430B Array | 1437665_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1437665_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 460113 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000460113 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2608406 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2608406 | - | - |
mouse-6 v1.1 (Illumina) | scl39112.5.1_8-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 460113 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl39112.5.1_8-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 460113 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2608406 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2608406 | - | - |
