Assay Details
Gene Symbol: Il22ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC124998.1 | - | 4 - 5 | 546 | 40044741 | 138 |
BC126872.1 | - | 4 - 5 | 556 | 40044756 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
