Assay Details
Gene Symbol: Grhl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013756.1 | NP_001013778.1 | 12 - 13 | 1651 | 65 | |
GenBank mRNA | BC089372.1 | - | 12 - 13 | 1651 | 30607679 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
