Assay Details
Gene Symbol: Irak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177973.1 | NP_001171444.1 | 2 - 3 | 481 | 97 | |
NM_001177974.1 | NP_001171445.1 | 2 - 3 | 481 | 97 | ||
NM_001177975.1 | NP_001171446.1 | 2 - 3 | 478 | 97 | ||
NM_001177976.1 | NP_001171447.1 | 2 - 3 | 481 | 97 | ||
NM_008363.2 | NP_032389.2 | 2 - 3 | 481 | 97 | ||
GenBank mRNA | AB088370.1 | - | 2 - 3 | 308 | 97 | |
AF103876.1 | - | 2 - 3 | 481 | 97 | ||
AK045958.1 | - | 2 - 3 | 387 | 97 | ||
AK052933.1 | - | 1 - 2 | 463 | 97 | ||
AK143694.1 | - | 2 - 3 | 389 | 97 | ||
AY184362.1 | - | 2 - 3 | 481 | 97 | ||
AY184363.1 | - | 2 - 3 | 481 | 97 | ||
AY184364.1 | - | 2 - 3 | 481 | 97 | ||
BC004778.1 | - | 2 - 3 | 473 | 3257579 | 97 | |
BY729815.1 | - | 2 - 3 | 480 | 96 | ||
U56773.2 | - | 2 - 3 | 305 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
