Assay Details
Gene Symbol: Irak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177973.1 | NP_001171444.1 | 5 - 6 | 907 | 101 | |
NM_001177974.1 | NP_001171445.1 | 5 - 6 | 904 | 101 | ||
NM_001177975.1 | NP_001171446.1 | 5 - 6 | 904 | 101 | ||
NM_001177976.1 | NP_001171447.1 | 5 - 6 | 904 | 101 | ||
NM_008363.2 | NP_032389.2 | 5 - 6 | 904 | 101 | ||
GenBank mRNA | AB088370.1 | - | 5 - 6 | 731 | 101 | |
AF103876.1 | - | 5 - 6 | 904 | 101 | ||
AK045958.1 | - | 5 - 6 | 813 | 101 | ||
AK052933.1 | - | 4 - 5 | 889 | 101 | ||
AK143694.1 | - | 5 - 6 | 815 | 101 | ||
AK144879.1 | - | 5 - 6 | 813 | 101 | ||
AK210148.1 | - | 1 - 2 | 171 | 101 | ||
AY184362.1 | - | 5 - 6 | 904 | 101 | ||
AY184363.1 | - | 5 - 6 | 904 | 101 | ||
AY184364.1 | - | 5 - 6 | 904 | 101 | ||
BC004778.1 | - | 5 - 6 | 899 | 3257579 | 101 | |
U56773.2 | - | 5 - 6 | 728 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
