Assay Details
Gene Symbol: Enpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF339910.1 | - | 12 - 13 | 1220 | 72 | |
AK029600.1 | - | 12 - 13 | 1132 | 72 | ||
AK036952.1 | - | 12 - 13 | 1229 | 72 | ||
AK041894.1 | - | 7 - 8 | 593 | 72 | ||
AK088857.1 | - | 12 - 13 | 1227 | 72 | ||
AK145233.1 | - | 12 - 13 | 1223 | 72 | ||
AK164619.1 | - | 12 - 13 | 1202 | 72 | ||
BC050011.1 | - | 12 - 13 | 1374 | 72 | ||
J02700.2 | - | 12 - 13 | 1331 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
