Assay Details
Gene Symbol: Slc6a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008135.4 | NP_032161.2 | 5 - 6 | 891 | 95 | |
GenBank mRNA | AK014572.1 | - | 735 | 95 | ||
AK048168.1 | - | 775 | 95 | |||
BC021828.1 | - | 5 - 6 | 714 | 4194951 | 95 | |
X67056.1 | - | 5 - 6 | 824 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
