Assay Details
Gene Symbol: Artn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284191.1 | NP_001271120.1 | 1 - 2 | 381 | 121 | |
NM_001284192.1 | NP_001271121.1 | 1 - 2 | 381 | 121 | ||
NM_001284193.1 | NP_001271122.1 | 1 - 2 | 381 | 121 | ||
GenBank mRNA | AK015393.1 | - | 1 - 2 | 89 | 121 | |
AK145190.1 | - | 1 - 2 | 89 | 121 | ||
AK156507.1 | - | 1 - 2 | 139 | 121 | ||
AK166013.1 | - | 1 - 2 | 118 | 121 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK015393.1, AK156507.1, NM_001284191.1, NM_001284192.1, NM_001284193.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
