Assay Details
Gene Symbol: Artn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284191.1 | NP_001271120.1 | 2 - 3 | 523 | 65 | |
NM_009711.4 | NP_033841.1 | 2 - 3 | 607 | 65 | ||
GenBank mRNA | AK015393.1 | - | 2 - 3 | 231 | 65 | |
AK053914.1 | - | 2 - 3 | 608 | 65 | ||
AK166013.1 | - | 2 - 3 | 260 | 65 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK015393.1, AK053914.1, NM_001284191.1, NM_009711.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
