Assay Details
Gene Symbol: Ascc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029291.1 | NP_083567.1 | 15 - 16 | 1729 | 55 | |
GenBank mRNA | AK148105.1 | - | 15 - 16 | 1761 | 55 | |
BC013537.1 | - | 15 - 16 | 1729 | 4240784 | 55 | |
BC024840.1 | - | 8 - 9 | 880 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
