Assay Details
Gene Symbol: Cdc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243769.1 | NP_001230698.1 | 4 - 5 | 453 | 74 | |
NM_009861.3 | NP_033991.1 | 4 - 5 | 453 | 74 | ||
GenBank mRNA | AK003098.1 | - | 4 - 5 | 374 | 74 | |
AK051543.1 | - | 4 - 5 | 374 | 74 | ||
AK075567.1 | - | 4 - 5 | 372 | 74 | ||
AK144216.1 | - | 4 - 5 | 376 | 74 | ||
AK145659.1 | - | 3 - 4 | 217 | 74 | ||
AK150563.1 | - | 4 - 5 | 386 | 74 | ||
AK151087.1 | - | 4 - 5 | 398 | 74 | ||
AK151726.1 | - | 4 - 5 | 375 | 74 | ||
AK153564.1 | - | 4 - 5 | 397 | 74 | ||
AK154870.1 | - | 4 - 5 | 372 | 74 | ||
AK159470.1 | - | 4 - 5 | 415 | 74 | ||
AK166281.1 | - | 4 - 5 | 372 | 74 | ||
AK167195.1 | - | 4 - 5 | 361 | 74 | ||
AK167609.1 | - | 4 - 5 | 371 | 74 | ||
AK168013.1 | - | 3 - 4 | 366 | 74 | ||
AK168076.1 | - | 4 - 5 | 366 | 74 | ||
AK168089.1 | - | 3 - 4 | 366 | 74 | ||
AK168276.1 | - | 4 - 5 | 366 | 74 | ||
AK168503.1 | - | 4 - 5 | 387 | 74 | ||
AK168758.1 | - | 4 - 5 | 406 | 74 | ||
AK168820.1 | - | 4 - 5 | 379 | 74 | ||
AK169122.1 | - | 4 - 5 | 387 | 74 | ||
AK169232.1 | - | 4 - 5 | 366 | 74 | ||
AK169805.1 | - | 4 - 5 | 391 | 74 | ||
AK213611.1 | - | 3 - 4 | 227 | 74 | ||
AK218916.1 | - | 1 - 2 | 84 | 74 | ||
BC064792.1 | - | 4 - 5 | 359 | 30105331 | 74 | |
CD551112.1 | - | 4 - 5 | 440 | 74 | ||
L11318.1 | - | 3 - 4 | 289 | 74 | ||
L78075.1 | - | 3 - 4 | 293 | 74 | ||
U37720.1 | - | 4 - 5 | 361 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
