Assay Details
Gene Symbol: Sgsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162965.1 | NP_001156437.1 | 5 - 6 | 544 | 79 | |
NM_001254731.1 | NP_001241660.1 | 5 - 6 | 544 | 79 | ||
NM_172718.3 | NP_766306.2 | 5 - 6 | 544 | 79 | ||
GenBank mRNA | AK044716.1 | - | 5 - 6 | 498 | 79 | |
AK053555.1 | - | 5 - 6 | 478 | 79 | ||
AK170021.1 | - | 5 - 6 | 536 | 79 | ||
BC138049.1 | - | 5 - 6 | 470 | 8861068 | 79 | |
BC138050.1 | - | 5 - 6 | 470 | 8861069 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
