Assay Details
Gene Symbol: Psd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028627.2 | NP_082903.2 | 11 - 12 | 2301 | 88 | |
GenBank mRNA | AK220438.1 | - | 6 - 7 | 2418 | 88 | |
BC043135.1 | - | 7 - 8 | 1130 | 88 | ||
BC057353.1 | - | 11 - 12 | 2302 | 88 | ||
BC138652.1 | - | 9 - 10 | 2160 | 8861672 | 88 | |
BC145352.1 | - | 9 - 10 | 2163 | 9053908 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
