Assay Details
Gene Symbol: Psd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028627.2 | NP_082903.2 | 7 - 8 | 1960 | 68 | |
GenBank mRNA | AK220438.1 | - | 2 - 3 | 2077 | 68 | |
BC043135.1 | - | 3 - 4 | 789 | 68 | ||
BC057353.1 | - | 7 - 8 | 1960 | 68 | ||
BC138652.1 | - | 5 - 6 | 1819 | 8861672 | 68 | |
BC145352.1 | - | 5 - 6 | 1822 | 9053908 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
