Assay Details
Gene Symbol: Wwc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133791.4 | NP_598552.2 | 11 - 12 | 2256 | 85 | |
GenBank mRNA | AB045323.1 | - | 11 - 12 | 2281 | 85 | |
AK032989.1 | - | 7 - 8 | 1322 | 85 | ||
AK079375.1 | - | 11 - 12 | 2092 | 85 | ||
AK147648.1 | - | 2250 | 85 | |||
BC054434.1 | - | 1 - 2 | 537 | 85 | ||
BC067050.1 | - | 11 - 12 | 2256 | 30537299 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
