Assay Details
Gene Symbol: Taf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081288.1 | NP_001074757.1 | 12 - 13 | 1788 | 138 | |
GenBank mRNA | AK147903.1 | - | 12 - 13 | 1790 | 138 | |
BC147638.1 | - | 12 - 13 | 1739 | 9056628 | 138 | |
BC151079.1 | - | 12 - 13 | 1739 | 9087992 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
