Assay Details
Gene Symbol: Taf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081288.1 | NP_001074757.1 | 18 - 19 | 2593 | 151 | |
GenBank mRNA | BC147638.1 | - | 18 - 19 | 2544 | 9056628 | 151 |
BC151079.1 | - | 18 - 19 | 2544 | 9087992 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
