Assay Details
Gene Symbol: Ppp1r32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005821.1 | - | 6 - 7 | 704 | 108 | |
BC118523.1 | - | 6 - 7 | 648 | 40105249 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
