Assay Details
Gene Symbol: Adgrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174991.3 | NP_778156.2 | 10 - 11 | 2832 | 62 | |
GenBank mRNA | AK147456.1 | - | 10 - 11 | 2817 | 62 | |
AK147459.1 | - | 10 - 11 | 2785 | 62 | ||
AK147494.1 | - | 10 - 11 | 2619 | 62 | ||
AK147600.1 | - | 10 - 11 | 2594 | 62 | ||
AK147607.1 | - | 10 - 11 | 2836 | 62 | ||
AK220481.1 | - | 10 - 11 | 2211 | 62 | ||
AY168408.1 | - | 10 - 11 | 2344 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
