Assay Details
Gene Symbol: Mtbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168250.1 | NP_001161722.1 | 8 - 9 | 1069 | 75 | |
NM_134092.3 | NP_598853.3 | 8 - 9 | 1069 | 75 | ||
GenBank mRNA | AJ278508.1 | - | 8 - 9 | 990 | 75 | |
AK080036.1 | - | 8 - 9 | 883 | 75 | ||
AK088549.1 | - | 8 - 9 | 883 | 75 | ||
AK133855.1 | - | 8 - 9 | 917 | 75 | ||
BC005500.1 | - | 1 - 2 | 66 | 75 | ||
BC052197.1 | - | 8 - 9 | 1069 | 6511752 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
