Assay Details
Gene Symbol: Fbxw8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172721.2 | NP_766309.2 | 7 - 8 | 1313 | 76 | |
GenBank mRNA | AK031815.1 | - | 7 - 8 | 1307 | 76 | |
AK034447.1 | - | 7 - 8 | 1241 | 76 | ||
AK047695.1 | - | 7 - 8 | 1281 | 76 | ||
AK047756.1 | - | 7 - 8 | 1255 | 76 | ||
AK053794.1 | - | 7 - 8 | 1314 | 76 | ||
AK134291.1 | - | 7 - 8 | 1320 | 76 | ||
AK214176.1 | - | 2 - 3 | 274 | 76 | ||
AK215899.1 | - | 2 - 3 | 275 | 76 | ||
BC009095.1 | - | 7 - 8 | 1266 | 3158218 | 76 | |
BC024091.1 | - | 1 - 2 | 189 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
