Assay Details
Gene Symbol: Fbxw8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172721.2 | NP_766309.2 | 8 - 9 | 1447 | 69 | |
GenBank mRNA | AK031815.1 | - | 8 - 9 | 1441 | 69 | |
AK034447.1 | - | 8 - 9 | 1375 | 69 | ||
AK053794.1 | - | 8 - 9 | 1448 | 69 | ||
AK134291.1 | - | 8 - 9 | 1454 | 69 | ||
BC009095.1 | - | 8 - 9 | 1400 | 3158218 | 69 | |
BC024091.1 | - | 2 - 3 | 323 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
