Assay Details
Gene Symbol: Tnks2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163635.1 | NP_001157107.1 | 13 - 14 | 1840 | 71 | |
GenBank mRNA | AK047094.1 | - | 13 - 14 | 1699 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
