Assay Details
Gene Symbol: Tnks2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163635.1 | NP_001157107.1 | 9 - 10 | 1408 | 150 | |
GenBank mRNA | AK047094.1 | - | 9 - 10 | 1267 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
