Assay Details
Gene Symbol: Helq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081107.1 | NP_001074576.1 | 13 - 14 | 2674 | 59 | |
GenBank mRNA | BC082601.1 | - | 13 - 14 | 2638 | 30662103 | 59 |
BC109169.2 | - | 13 - 14 | 2542 | 59 | ||
BC109170.2 | - | 13 - 14 | 2547 | 40048258 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
