Assay Details
Gene Symbol: Incenp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016692.3 | NP_057901.2 | 14 - 15 | 2302 | 174 | |
GenBank mRNA | AB100432.1 | - | 13 - 14 | 2096 | 174 | |
AB100433.1 | - | 14 - 15 | 2108 | 174 | ||
AF117610.1 | - | 14 - 15 | 2238 | 174 | ||
AK034764.1 | - | 13 - 14 | 2562 | 174 | ||
AK081841.1 | - | 14 - 15 | 2236 | 174 | ||
AK088627.1 | - | 14 - 15 | 2259 | 174 | ||
BC052414.1 | - | 14 - 15 | 2235 | 6401188 | 174 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
