Assay Details
Gene Symbol: Incenp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016692.3 | NP_057901.2 | 9 - 10 | 1595 | 99 | |
GenBank mRNA | AB100432.1 | - | 8 - 9 | 1389 | 99 | |
AB100433.1 | - | 8 - 9 | 1389 | 99 | ||
AK034764.1 | - | 9 - 10 | 1524 | 99 | ||
AK081841.1 | - | 9 - 10 | 1529 | 99 | ||
AK088627.1 | - | 9 - 10 | 1552 | 99 | ||
BC052414.1 | - | 9 - 10 | 1528 | 6401188 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
