Assay Details
Gene Symbol: Mapkapk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010765.2 | NP_034895.1 | 5 - 6 | 1089 | 71 | |
GenBank mRNA | AF039840.1 | - | 5 - 6 | 1103 | 71 | |
AY533679.1 | - | 5 - 6 | 397 | 71 | ||
AY533680.2 | - | 5 - 6 | 397 | 71 | ||
BC019184.1 | - | 5 - 6 | 511 | 5010305 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
