Assay Details
Gene Symbol: Loxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF338440.1 | - | 9 - 10 | 1593 | 57 | |
BC068307.1 | - | 10 - 11 | 1640 | 30059745 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
