Assay Details
Gene Symbol: Dgkq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199011.1 | NP_950176.1 | 21 - 22 | 2625 | 59 | |
GenBank mRNA | AK043933.1 | - | 2792 | 59 | ||
AK048764.1 | - | 20 - 21 | 2943 | 59 | ||
AK077288.1 | - | 1 - 2 | 73 | 59 | ||
AK132151.1 | - | 20 - 21 | 2532 | 59 | ||
AK134745.1 | - | 8 - 9 | 1688 | 59 | ||
AK170840.1 | - | 18 - 19 | 2591 | 59 | ||
BC062929.1 | - | 21 - 22 | 2625 | 6855058 | 59 | |
BC066012.1 | - | 19 - 20 | 3054 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
