Assay Details
Gene Symbol: Dgkq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199011.1 | NP_950176.1 | 5 - 6 | 720 | 87 | |
GenBank mRNA | AK043933.1 | - | 570 | 87 | ||
AK048764.1 | - | 5 - 6 | 762 | 87 | ||
AK132151.1 | - | 5 - 6 | 717 | 87 | ||
AK170840.1 | - | 5 - 6 | 741 | 87 | ||
BC062929.1 | - | 5 - 6 | 720 | 6855058 | 87 | |
BC066012.1 | - | 5 - 6 | 697 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
