Assay Details
Gene Symbol: Sort1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271599.1 | NP_001258528.1 | 10 - 11 | 1334 | 95 | |
NM_019972.3 | NP_064356.2 | 10 - 11 | 1334 | 95 | ||
GenBank mRNA | AF175279.1 | - | 10 - 11 | 1263 | 95 | |
AK147442.1 | - | 10 - 11 | 1317 | 95 | ||
BC034129.1 | - | 7 - 8 | 767 | 95 | ||
BC056343.1 | - | 10 - 11 | 1266 | 5704346 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
