Assay Details
Gene Symbol: Lig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083188.1 | NP_001076657.1 | 7 - 8 | 835 | 64 | |
NM_001199310.1 | NP_001186239.1 | 7 - 8 | 843 | 64 | ||
NM_010715.2 | NP_034845.2 | 7 - 8 | 760 | 64 | ||
GenBank mRNA | AK153993.1 | - | 7 - 8 | 722 | 64 | |
AK168643.1 | - | 7 - 8 | 837 | 64 | ||
BC028287.1 | - | 7 - 8 | 685 | 4911261 | 64 | |
BC138541.1 | - | 7 - 8 | 843 | 8861561 | 64 | |
BC138542.1 | - | 7 - 8 | 843 | 8861562 | 64 | |
U04674.1 | - | 6 - 7 | 628 | 64 | ||
U19604.1 | - | 7 - 8 | 873 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
