Assay Details
Gene Symbol: Lig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083188.1 | NP_001076657.1 | 8 - 9 | 964 | 76 | |
NM_001199310.1 | NP_001186239.1 | 8 - 9 | 972 | 76 | ||
NM_010715.2 | NP_034845.2 | 8 - 9 | 889 | 76 | ||
GenBank mRNA | AK153993.1 | - | 8 - 9 | 851 | 76 | |
AK161793.1 | - | 1 - 2 | 47 | 76 | ||
AK168643.1 | - | 8 - 9 | 966 | 76 | ||
BC028287.1 | - | 8 - 9 | 814 | 4911261 | 76 | |
BC138541.1 | - | 8 - 9 | 972 | 8861561 | 76 | |
BC138542.1 | - | 8 - 9 | 972 | 8861562 | 76 | |
U04674.1 | - | 7 - 8 | 757 | 76 | ||
U19604.1 | - | 8 - 9 | 1002 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
