Assay Details
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130409.1 | NP_001123881.1 | 19 - 20 | 1986 | 76 | |
NM_007982.2 | NP_032008.2 | 18 - 19 | 1831 | 76 | ||
GenBank mRNA | AB011499.1 | - | 19 - 20 | 1642 | 76 | |
AK079821.1 | - | 19 - 20 | 2083 | 76 | ||
AK220543.1 | - | 18 - 19 | 1831 | 76 | ||
BC030180.1 | - | 19 - 20 | 1916 | 76 | ||
JN971016.1 | - | 18 - 19 | 1558 | 76 | ||
M95408.1 | - | 17 - 18 | 1628 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
