Assay Details
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130409.1 | NP_001123881.1 | 27 - 28 | 2963 | 63 | |
NM_007982.2 | NP_032008.2 | 26 - 27 | 2808 | 63 | ||
GenBank mRNA | AB011499.1 | - | 27 - 28 | 2619 | 63 | |
AB030035.1 | - | 24 - 25 | 2229 | 63 | ||
AK079821.1 | - | 27 - 28 | 3060 | 63 | ||
AK220543.1 | - | 26 - 27 | 2808 | 63 | ||
BC030180.1 | - | 27 - 28 | 2893 | 63 | ||
JN971016.1 | - | 26 - 27 | 2535 | 63 | ||
M95408.1 | - | 25 - 26 | 2605 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
