Assay Details
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130409.1 | NP_001123881.1 | 9 - 10 | 1114 | 131 | |
NM_007982.2 | NP_032008.2 | 8 - 9 | 959 | 131 | ||
GenBank mRNA | AB011499.1 | - | 6 - 7 | 647 | 131 | |
AB030035.1 | - | 6 - 7 | 647 | 131 | ||
AK220543.1 | - | 8 - 9 | 959 | 131 | ||
BC030180.1 | - | 9 - 10 | 1044 | 131 | ||
JN971016.1 | - | 6 - 7 | 647 | 131 | ||
M95408.1 | - | 7 - 8 | 756 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
