Assay Details
Gene Symbol: Chst12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021528.3 | NP_067503.3 | 2 | 1623 | 93 | |
GenBank mRNA | AJ289132.1 | - | 1 | 1536 | 93 | |
AK142155.1 | - | 2 | 1621 | 93 | ||
AK163588.1 | - | 2 | 1595 | 93 | ||
AK170484.1 | - | 2 | 1590 | 93 | ||
AK171208.1 | - | 2 | 1590 | 93 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141810 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141719 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141718 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141812 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141816 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141811 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141720 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141722 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141721 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141815 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464023 | AK171208.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464023 | BC003201.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464023 | AK163588.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464023 | AK170484.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464023 | AJ289132.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464023 | AK171208.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464023 | BC003201.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464023 | AK163588.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464023 | AK170484.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P464023 | AJ289132.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 943546 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 276149 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 129713 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555209 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 288850 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467161 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 755202 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 41910 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 241809 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293436 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621309 | AF051347.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621309 | AF097438.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621309 | AK003429.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621309 | BC058805.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621309 | BC089464.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621309 | NM_009052.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 279686 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644517 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 134702 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129908 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164794 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464023 | AK171208.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464023 | BC003201.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464023 | AK163588.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464023 | AK170484.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464023 | AJ289132.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464023 | AK171208.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464023 | BC003201.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464023 | AK163588.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464023 | AK170484.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P464023 | AJ289132.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P176409 | BC003201.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464023 | AK171208.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464023 | BC003201.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464023 | AK163588.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464023 | AK170484.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464023 | AJ289132.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464023 | AK171208.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464023 | BC003201.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464023 | AK163588.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464023 | AK170484.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P464023 | AJ289132.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141707 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141734 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141705 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141709 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141725 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141722 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141714 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141735 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141726 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141730 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141716 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141737 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141717 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141733 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141727 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141711 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141720 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24141724 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129908 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164794 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01236496 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01236489 | - | - |
Whole Mouse Genome G4122A | A_52_P176409 | BC003201.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464023 | AK171208.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464023 | BC003201.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464023 | AK163588.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464023 | AK170484.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464023 | AJ289132.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464023 | AK171208.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464023 | BC003201.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464023 | AK163588.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464023 | AK170484.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464023 | AJ289132.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129282 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129908 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164794 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04962238 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04962234 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04250617 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141811 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141812 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141816 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141815 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141718 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141810 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141722 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141719 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141721 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24141720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24141737 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04603333 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141724 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141714 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04962238 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141750 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04469238 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141740 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141728 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141707 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04469237 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04133590 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04384296 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04333653 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04873720 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04250614 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04250616 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141712 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141722 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141708 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141738 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04469230 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141725 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141717 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141704 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04227382 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141711 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04133593 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04469234 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141742 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04457236 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04962235 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04628460 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141733 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04133594 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24141709 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10527011 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10527010 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448477_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10527009 | - | - |
Mouse Genome 430 2.0 Array | 1448477_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1448477_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 59031_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 59031_at | - | - |
Mouse Expression Set 430 | 1448477_PM_at | - | - |
Mouse Expression Set 430 | 1448477_at | - | - |
Mouse Genome 430A 2.0 Array | 1448477_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1448477_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448477_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448477_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 686778 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2030438 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2030438 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002030438 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2710205 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2710205 | - | - |
mouse-6 v1.1 (Illumina) | scl27049.4_88-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2030438 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl27049.4_88-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2030438 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2710205 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2710205 | - | - |
