Assay Details
Gene Symbol: Asphd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028386.1 | NP_082662.1 | 2 - 3 | 1176 | 83 | |
GenBank mRNA | AK013495.1 | - | 2 - 3 | 428 | 83 | |
AK020319.1 | - | 2 - 3 | 259 | 83 | ||
BC046606.1 | - | 2 - 3 | 1176 | 6314853 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
peptidyl-amino acid modification
sulfonate dioxygenase activity
molecular_function
oxidoreductase activity
2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
procollagen-proline dioxygenase activity
hypophosphite dioxygenase activity
DNA-N1-methyladenine dioxygenase activity
metal ion binding
dioxygenase activity
C-19 gibberellin 2-beta-dioxygenase activity
C-20 gibberellin 2-beta-dioxygenase activity
biological_process
peptidyl-amino acid modification
sulfonate dioxygenase activity
molecular_function
oxidoreductase activity
2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
procollagen-proline dioxygenase activity
hypophosphite dioxygenase activity
DNA-N1-methyladenine dioxygenase activity
metal ion binding
dioxygenase activity
C-19 gibberellin 2-beta-dioxygenase activity
C-20 gibberellin 2-beta-dioxygenase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23991928 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23991929 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23991927 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285779 | BC046606.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285779 | NM_028386.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285779 | AK013495.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285779 | AK020319.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285779 | BC046606.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285779 | NM_028386.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285779 | AK013495.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P285779 | AK020319.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 660968 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883440 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362768 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52442 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126395 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285779 | BC046606.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285779 | NM_028386.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285779 | AK013495.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285779 | AK020319.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285779 | BC046606.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285779 | NM_028386.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285779 | AK013495.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P285779 | AK020319.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991906 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991915 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991942 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991920 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991933 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991925 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991895 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991902 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991927 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991943 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991940 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991919 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991916 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991922 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991930 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991948 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991928 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991893 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991938 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991936 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991898 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991903 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991899 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991937 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991896 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991945 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991931 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991911 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991894 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991941 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991914 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991918 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991908 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991917 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23991924 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126395 | - | - |
Whole Mouse Genome G4122A | A_51_P285779 | BC046606.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285779 | NM_028386.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285779 | AK013495.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285779 | AK020319.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285779 | BC046606.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285779 | NM_028386.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285779 | AK013495.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285779 | AK020319.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126395 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04726225 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04726251 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04726227 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04726235 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04726215 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04726248 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23991928 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23991929 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23991927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991963 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991884 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991881 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23991847 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991894 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991853 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991883 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991918 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991920 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04726245 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991931 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991902 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991896 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991869 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991862 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991938 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04272492 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04496147 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991848 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04726243 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04614704 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991916 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04902123 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991851 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991925 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991866 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04986062 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991873 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04234296 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04173530 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04726220 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991842 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04726227 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991936 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04726239 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991951 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991865 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991955 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991943 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991864 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991949 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04725558 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991963 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04468787 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991850 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04180963 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991945 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991845 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991940 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991871 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991906 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04829114 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991859 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991953 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04656543 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991960 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04468783 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991857 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991933 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04468775 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991886 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991944 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991899 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991898 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991847 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991843 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04197728 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991884 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04234294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991922 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991942 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991893 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991937 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991852 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991867 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991890 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991903 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23991908 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10532539 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532540 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532541 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452958_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10532538 | - | - |
Mouse Genome 430 2.0 Array | 1452958_at | - | - |
Mouse Genome 430 2.0 Array | 1435845_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1452958_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1435845_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72898_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72898_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500026773.mm.1 | TC0500003018.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500026773.mm.1 | NM_028386.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500026773.mm.1 | NM_028386.1 | Refseq |
Mouse Expression Set 430 | 1452958_PM_at | - | - |
Mouse Expression Set 430 | 1452958_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1452958_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1452958_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 919735 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3162247 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_3162247 | - | - |
mouse-6 v1.1 (Illumina) | GI_55926228-S | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3162247 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3162247 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | GI_55926228-S | - | - |
