Assay Details
Gene Symbol: Naa25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172722.3 | NP_766310.2 | 20 - 21 | 2470 | 124 | |
GenBank mRNA | AK041906.1 | - | 6 - 7 | 787 | 124 | |
AK049325.1 | - | 20 - 21 | 2471 | 124 | ||
AK051815.1 | - | 3 - 4 | 226 | 124 | ||
AK149758.1 | - | 20 - 21 | 2471 | 124 | ||
AK165175.1 | - | 20 - 21 | 2462 | 124 | ||
AK167405.1 | - | 20 - 21 | 2488 | 124 | ||
AK167491.1 | - | 20 - 21 | 2488 | 124 | ||
BC028868.1 | - | 18 - 19 | 2250 | 124 | ||
BC037595.1 | - | 20 - 21 | 2451 | 124 | ||
BC055480.1 | - | 19 - 20 | 2372 | 124 | ||
BC082314.1 | - | 20 - 21 | 2456 | 124 | ||
BC106182.1 | - | 20 - 21 | 2445 | 124 | ||
BC132172.1 | - | 20 - 21 | 2468 | 40130449 | 124 | |
BC138093.1 | - | 20 - 21 | 2468 | 8861112 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
