Assay Details
Gene Symbol: P2rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164834.1 | NP_001158306.1 | 11 - 12 | 1233 | 120 | |
GenBank mRNA | AB094663.1 | - | 11 - 12 | 1153 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
