Assay Details
Gene Symbol: Ush1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163733.1 | NP_001157205.1 | 5 - 6 | 517 | 61 | |
NM_001291182.1 | NP_001278111.1 | 5 - 6 | 517 | 61 | ||
NM_023649.2 | NP_076138.2 | 5 - 6 | 517 | 61 | ||
NM_153677.2 | NP_710143.2 | 5 - 6 | 517 | 61 | ||
GenBank mRNA | AF228924.1 | - | 5 - 6 | 517 | 61 | |
AF228925.1 | - | 5 - 6 | 517 | 61 | ||
AY103465.1 | - | 5 - 6 | 517 | 61 | ||
AY246556.1 | - | 5 - 6 | 491 | 61 | ||
BC010819.1 | - | 5 - 6 | 519 | 4217086 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
