Assay Details
Gene Symbol: Abcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018795.2 | NP_061265.2 | 7 - 8 | 821 | 79 | |
GenBank mRNA | AB028737.1 | - | 7 - 8 | 827 | 79 | |
BC040400.1 | - | 7 - 8 | 829 | 79 | ||
EF109740.1 | - | 7 - 8 | 821 | 79 | ||
EF109741.1 | - | 7 - 8 | 821 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
