Assay Details
Gene Symbol: Oas1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145210.2 | NP_660211.2 | 5 - 6 | 1088 | 71 | |
GenBank mRNA | AB067531.1 | - | 5 - 6 | 1062 | 71 | |
AK087823.1 | - | 5 - 6 | 1088 | 71 | ||
AY055831.1 | - | 5 - 6 | 1061 | 71 | ||
BC053721.1 | - | 5 - 6 | 1085 | 30053184 | 71 | |
BC094918.1 | - | 5 - 6 | 1078 | 30050168 | 71 | |
BC108956.1 | - | 5 - 6 | 1084 | 40046020 | 71 | |
DV662831.1 | - | 1 - 2 | 121 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
