Assay Details
Gene Symbol: Zfp644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026856.2 | NP_081132.2 | 4 - 5 | 3296 | 69 | |
GenBank mRNA | AK122471.1 | - | 1 - 2 | 2869 | 69 | |
BC030844.1 | - | 2 - 3 | 941 | 69 | ||
BC057405.1 | - | 4 - 5 | 3174 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
