Assay Details
Gene Symbol: Zfp644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026856.2 | NP_081132.2 | 5 - 6 | 3904 | 92 | |
GenBank mRNA | AK122471.1 | - | 2 - 3 | 3477 | 92 | |
BC057405.1 | - | 5 - 6 | 3782 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
