Assay Details
Gene Symbol: Zfp644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026856.2 | NP_081132.2 | 2 - 3 | 359 | 140 | |
GenBank mRNA | AK053146.1 | - | 2 - 3 | 360 | 140 | |
BB663850.1 | - | 2 - 3 | 360 | 140 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK053146.1, NM_026856.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
