Assay Details
Gene Symbol: Gm6602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045362.1 | 2 - 3 | 489 | 184 | ||
GenBank mRNA | AK090313.1 | - | 2 - 3 | 490 | 184 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein arginine N-methyltransferase 6 | Prmt6 |
AW124876, BB233495, Hrmt1l6 |
-1.7mb | Anti-sense | Chr. 3 - 110246104 - 110250999 on NCBI Build GRCm38 |
Downstream | amylase 2b | Amy2b |
Amy-X, Amy2-2, OTTMUSG00000022462, mAmy2-1 |
1.1mb | Anti-sense | Chr. 3 - 113147656 - 113156727 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121012 | AK090313.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136237 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883716 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426921 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 461252 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149887 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662546 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 468373 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 961829 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380455 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 460517 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 920534 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601255 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 799723 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 272721 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918089 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 103481 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 755178 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 131099 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108265 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411190 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583745 | AK030108.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 776294 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174489 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155114 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581156 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 241432 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 41136 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323077 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267265 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822625 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 165691 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562542 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316507 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 387700 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45276 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444070 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 459214 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551464 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717597 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 232840 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 732012 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218634 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 247081 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884286 | AK131666.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884286 | AK151792.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884286 | AK169002.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884286 | BC006762.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884286 | L37296.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884286 | NM_007522.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 176769 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 86269 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 169610 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 167674 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92426 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127710 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139095 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 106231 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10007 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121012 | AK090313.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05032548 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05032440 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05032517 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05032400 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05032366 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05032423 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05032526 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05032385 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05032490 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05032455 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05032462 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05032582 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121012 | AK090313.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05032517 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05032582 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05032455 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05032366 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05032423 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429575 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429552 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429883 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429774 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429866 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429573 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429871 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429864 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429759 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429880 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429571 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429884 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02366981 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429566 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429769 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429777 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429876 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429577 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429765 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429567 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429772 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429770 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02299632 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429556 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429763 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429758 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429561 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429868 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429767 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429861 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429874 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429776 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429569 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02767047 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429570 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429886 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429877 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429885 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429761 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429873 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429572 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429554 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429870 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22429860 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02549615 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0300009553.mm.1 | TC0300001167.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300009553.mm.1 | NR_045362.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300009553.mm.1 | NR_045362.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300009552.mm.1 | TC0300001167.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300009552.mm.1 | NR_045362.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300009552.mm.1 | NR_045362.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 335143 | - | - |
