Assay Details
Gene Symbol: Kif11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010615.1 | NP_034745.1 | 12 - 13 | 1685 | 70 | |
GenBank mRNA | AJ223293.1 | - | 11 - 12 | 1380 | 70 | |
AK050778.1 | - | 1 - 2 | 1472 | 70 | ||
AK083324.1 | - | 12 - 13 | 1662 | 70 | ||
AK147754.1 | - | 12 - 13 | 1709 | 70 | ||
BC060670.1 | - | 12 - 13 | 1685 | 6406754 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
